Search
Search

Efcab12| Gene Symbol: Efcab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749810.3 | XP_003749858.1 | 4 - 5 | 1940 | 67 | |
| XM_008763265.1 | XP_008761487.1 | 4 - 5 | 836 | 67 | ||
Set Membership: |
|