Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01459805_m1
          See other ETV6 GE Assays ›
          Gene Symbol:
          Etv6
          Gene Name:
          ets variant 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 167754684 - 167992219 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01459805_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Etv6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037353.1 NP_001032430.1 7 - 8 1266 66
          GenBank mRNA BC105773.1 - 7 - 8 1266 7930490 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001914.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Etv6

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147708 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 902 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13779 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99773 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153621 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46517 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74626 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129718 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033872 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033872 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033872 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P424396 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34115 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4160 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82047 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104947 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 237 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70720 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78446 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103691 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57983 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38508 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57535 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57297 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28550 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34815 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859234 - -
          Affymetrix Rat Gene 1.0 ST Array 10859230 - -
          Affymetrix Rat Gene 1.0 ST Array 10859228 - -
          Affymetrix Rat Gene 1.0 ST Array 10859236 - -
          Affymetrix Rat Gene 1.0 ST Array 10859231 - -
          Affymetrix Rat Gene 1.0 ST Array 10859233 - -
          Affymetrix Rat Gene 1.0 ST Array 10859232 - -
          Affymetrix Rat Gene 1.0 ST Array 10859235 - -
          Affymetrix Rat Gene 1.0 ST Array 10859229 - -
          Affymetrix Rat Gene 1.1 ST Array 10859227 - -
          Rat Genome 230 2.0 Array 1389938_at - -
          Rat Genome 230 2.0 Array 1385294_PM_at - -
          Rat Genome 230 2.0 Array 1385294_at - -
          Rat Genome 230 2.0 Array 1389938_PM_at - -
          Affymetrix HT Rat Focus Array 1385294_at - -
          Affymetrix HT Rat Focus Array 1385294_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400013841.rn.1 TC0400001914.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013843.rn.1 TC0400001914.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013844.rn.1 TC0400001914.rn.1 RTA TC
          Rat Expression Set 230 1389938_PM_at - -
          Rat Expression Set 230 1389938_at - -
          Rat Expression Set 230 1385294_at - -
          Rat Expression Set 230 1385294_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859227 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline