Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460107_m1
          See other NSRP1 GE Assays ›
          Gene Symbol:
          Nsrp1
          Gene Name:
          nuclear speckle splicing regulatory protein 1
          Gene Aliases:
          Ccdc55, NSrp70, RGD1309863
          Chromosome Location:
          Chr.10: 63098115 - 63133188 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460107_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nsrp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037189.1 NP_001032266.1 6 - 7 622 76
          XM_008768018.1 XP_008766240.1 5 - 6 1770 76
          GenBank mRNA BC099120.1 - 6 - 7 622 7462721 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001090.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nsrp1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA processing
          nucleocytoplasmic transport
          RNA splicing
          developmental process
          mRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1031638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104797 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1031638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1031638 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1031638 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1031638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50961 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12315 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10736480 - -
          Affymetrix Rat Gene 1.0 ST Array 10736481 - -
          Affymetrix Rat Gene 1.0 ST Array 10736477 - -
          Affymetrix Rat Gene 1.0 ST Array 10736482 - -
          Affymetrix Rat Gene 1.0 ST Array 10736479 - -
          Affymetrix Rat Gene 1.0 ST Array 10736478 - -
          Affymetrix Rat Gene 1.0 ST Array 10736483 - -
          Affymetrix Rat Gene 1.1 ST Array 10736476 - -
          Rat Genome 230 2.0 Array 1394794_PM_at - -
          Rat Genome 230 2.0 Array 1394794_at - -
          Rat Genome 230 2.0 Array 1382399_at - -
          Rat Genome 230 2.0 Array 1382904_PM_at - -
          Rat Genome 230 2.0 Array 1382904_at - -
          Rat Genome 230 2.0 Array 1382399_PM_at - -
          Affymetrix HT Rat Focus Array 1382904_at - -
          Affymetrix HT Rat Focus Array 1382904_PM_at - -
          Affymetrix HT Rat Focus Array 1382399_PM_at - -
          Affymetrix HT Rat Focus Array 1394794_PM_at - -
          Affymetrix HT Rat Focus Array 1382399_at - -
          Affymetrix HT Rat Focus Array 1394794_at - -
          Affymetrix Rat Clariom S Assay PSR1000009869.rn.1 TC1000001090.rn.1 RTA TC
          Rat Expression Set 230 1394794_PM_at - -
          Rat Expression Set 230 1382399_PM_at - -
          Rat Expression Set 230 1382904_at - -
          Rat Expression Set 230 1382904_PM_at - -
          Rat Expression Set 230 1382399_at - -
          Rat Expression Set 230 1394794_at - -
          Affymetrix Rat Gene 1.0 ST Array 10736476 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22423705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline