Search
Search
| Gene Symbol: Flii | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008279.1 | NP_001008280.1 | 18 - 19 | 2219 | 60 | |
| XM_006246468.2 | XP_006246530.1 | 18 - 19 | 2267 | 60 | ||
| GenBank mRNA | BC085829.1 | - | 18 - 19 | 2219 | 7133081 | 60 |
Set Membership: |
|