Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750953.3 | XP_003751001.1 | 7 - 8 | 1443 | 61 | |
XM_003750954.3 | XP_003751002.1 | 7 - 8 | 1046 | 61 | ||
XM_006247762.2 | XP_006247824.1 | 7 - 8 | 1046 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |