Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009962.1 | NP_001009962.1 | 2 - 3 | 513 | 63 | |
GenBank mRNA | AY800384.1 | - | 2 - 3 | 513 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |