Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460556_g1
          See other RGD1308134 GE Assays ›
          Gene Symbol:
          RGD1308134
          Gene Name:
          similar to RIKEN cDNA 1110020A23
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 56843846 - 56848270 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460556_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1308134
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127521.2 NP_001120993.1 4 - 5 459 121
          XM_006246652.2 XP_006246714.1 4 - 5 627 121
          XM_006246653.2 XP_006246715.1 3 - 4 525 121
          XM_006246655.2 XP_006246717.1 3 - 4 525 121
          GenBank mRNA BC158879.1 - 4 - 5 459 7936013 121
          FQ228078.1 - 1 - 2 155 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003121.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Process(es)

          DNA binding
          chromatin binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P204094 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P108724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329362 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P204094 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P204094 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P204094 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P108724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744386 - -
          Affymetrix Rat Gene 1.0 ST Array 10744388 - -
          Affymetrix Rat Gene 1.0 ST Array 10744387 - -
          Affymetrix Rat Gene 1.0 ST Array 10744389 - -
          Affymetrix Rat Gene 1.1 ST Array 10744385 - -
          Rat Genome 230 2.0 Array 1388665_PM_at - -
          Rat Genome 230 2.0 Array 1388665_at - -
          Affymetrix HT Rat Focus Array 1388665_PM_at - -
          Affymetrix HT Rat Focus Array 1388665_at - -
          Affymetrix Rat Clariom S Assay PSR1000027996.rn.1 TC1000003121.rn.1 RTA TC
          Rat Expression Set 230 1388665_PM_at - -
          Rat Expression Set 230 1388665_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744385 - -
          Rat Genome U34 Set rc_AA866371_s_at - -
          Rat Genome U34 Set rc_AA946448_at - -
          Rat Genome U34 Set rc_AI229695_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368376 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline