Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014227.1 | NP_001014249.1 | 3 - 4 | 297 | 73 | |
GenBank mRNA | BC088458.1 | - | 3 - 4 | 297 | 7127323 | 73 |
FQ210234.1 | - | 3 - 4 | 315 | 73 | ||
FQ230415.1 | - | 3 - 4 | 273 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |