Search
Search

LOC100158225| Gene Symbol: LOC100158225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127606.1 | NP_001121078.1 | 7 - 8 | 871 | 64 | |
| GenBank mRNA | BC166509.1 | - | 7 - 8 | 871 | 7127250 | 64 |
Set Membership: |
|