Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108279.1 | NP_001101749.1 | 3 - 4 | 774 | 130 | |
XM_008767841.1 | XP_008766063.1 | 4 - 5 | 869 | 130 | ||
XM_008767842.1 | XP_008766064.1 | 4 - 5 | 1159 | 130 | ||
XM_008767843.1 | XP_008766065.1 | 4 - 5 | 859 | 130 | ||
GenBank mRNA | FQ223808.1 | - | 4 - 5 | 830 | 130 | |
FQ224714.1 | - | 4 - 5 | 830 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |