Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460731_m1
          See other SLC47A1 GE Assays ›
          Gene Symbol:
          Slc47a1
          Gene Name:
          solute carrier family 47 (multidrug and toxin extrusion), member 1
          Gene Aliases:
          MATE1, RGD1311123
          Chromosome Location:
          Chr.10: 47632352 - 47666876 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460731_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc47a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014118.2 NP_001014140.1 14 - 15 1309 66
          GenBank mRNA AB248823.1 - 14 - 15 1309 66
          AB248824.1 - 14 - 15 1309 66
          BC088413.1 - 1329 7109897 66
          FQ217802.1 - 1323 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002950.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc47a1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          drug transmembrane transport
          organic cation transport
          drug transport
          hydrogen ion transmembrane transport
          monovalent cation:proton antiporter activity
          drug transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P131721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131721 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131721 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P131721 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19475 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10743403 - -
          Affymetrix Rat Gene 1.0 ST Array 10743404 - -
          Affymetrix Rat Gene 1.0 ST Array 10743393 - -
          Affymetrix Rat Gene 1.0 ST Array 10743389 - -
          Affymetrix Rat Gene 1.0 ST Array 10743398 - -
          Affymetrix Rat Gene 1.0 ST Array 10743392 - -
          Affymetrix Rat Gene 1.0 ST Array 10743408 - -
          Affymetrix Rat Gene 1.0 ST Array 10743395 - -
          Affymetrix Rat Gene 1.0 ST Array 10743399 - -
          Affymetrix Rat Gene 1.0 ST Array 10743394 - -
          Affymetrix Rat Gene 1.0 ST Array 10743407 - -
          Affymetrix Rat Gene 1.0 ST Array 10743391 - -
          Affymetrix Rat Gene 1.0 ST Array 10743388 - -
          Affymetrix Rat Gene 1.0 ST Array 10743397 - -
          Affymetrix Rat Gene 1.0 ST Array 10743390 - -
          Affymetrix Rat Gene 1.0 ST Array 10743405 - -
          Affymetrix Rat Gene 1.0 ST Array 10743396 - -
          Affymetrix Rat Gene 1.0 ST Array 10743406 - -
          Affymetrix Rat Gene 1.0 ST Array 10743400 - -
          Affymetrix Rat Gene 1.0 ST Array 10743401 - -
          Affymetrix Rat Gene 1.1 ST Array 10743387 - -
          Rat Genome 230 2.0 Array 1390835_at - -
          Rat Genome 230 2.0 Array 1390835_PM_at - -
          Affymetrix HT Rat Focus Array 1390835_PM_at - -
          Affymetrix HT Rat Focus Array 1390835_at - -
          Affymetrix Rat Clariom S Assay PSR1000025989.rn.1 TC1000002950.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000025988.rn.1 TC1000002950.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000025990.rn.1 TC1000002950.rn.1 RTA TC
          Rat Expression Set 230 1390835_at - -
          Rat Expression Set 230 1390835_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10743387 - -
          Rat Genome U34 Set rc_AI013568_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21079170 - -
          ABI Rat Genome Survey Microarray 22339991 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline