Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460737_m1
          See other THG1L GE Assays ›
          Gene Symbol:
          Thg1l
          Gene Name:
          tRNA-histidine guanylyltransferase 1-like (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 31044885 - 31052092 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460737_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thg1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013966.1 NP_001013988.1 5 - 6 752 103
          GenBank mRNA BC087596.1 - 5 - 6 752 7310343 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002639.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thg1l

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA modification
          tRNA processing
          protein homotetramerization
          magnesium ion binding
          ATP binding
          GTP binding
          tRNA guanylyltransferase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052230 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052230 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052230 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P173622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87100 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742242 - -
          Affymetrix Rat Gene 1.0 ST Array 10742237 - -
          Affymetrix Rat Gene 1.0 ST Array 10742240 - -
          Affymetrix Rat Gene 1.0 ST Array 10742241 - -
          Affymetrix Rat Gene 1.0 ST Array 10742238 - -
          Affymetrix Rat Gene 1.0 ST Array 10742239 - -
          Affymetrix Rat Gene 1.1 ST Array 10742236 - -
          Rat Genome 230 2.0 Array 1384330_at - -
          Rat Genome 230 2.0 Array 1384330_PM_at - -
          Affymetrix HT Rat Focus Array 1384330_at - -
          Affymetrix HT Rat Focus Array 1384330_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000023467.rn.1 TC1000002639.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000023468.rn.1 TC1000002639.rn.1 RTA TC
          Rat Expression Set 230 1384330_PM_at - -
          Rat Expression Set 230 1384330_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742236 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22072402 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline