Search
Search

Rab11fip3| Gene Symbol: Rab11fip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002727755.3 | XP_002727801.1 | 3 - 4 | 1913 | 101 | |
| XM_006246071.1 | XP_006246133.1 | 3 - 4 | 1913 | 101 | ||
| XM_220262.6 | XP_220262.5 | 3 - 4 | 1913 | 101 | ||
Set Membership: |
|