Search
Search
| Gene Symbol: Anxa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024156.2 | NP_077070.2 | 2 - 3 | 131 | 65 | |
| XM_006246340.2 | XP_006246402.1 | 2 - 3 | 869 | 65 | ||
| XM_006246341.2 | XP_006246403.1 | 2 - 3 | 870 | 65 | ||
| GenBank mRNA | BC072523.1 | - | 2 - 3 | 80 | 7105847 | 65 |
| CB585592.1 | - | 2 - 3 | 149 | 65 | ||
Set Membership: |
|