Search
Search
| Gene Symbol: Cpeb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106992.1 | NP_001100462.1 | 4 - 5 | 1426 | 69 | |
| XM_006245978.2 | XP_006246040.1 | 5 - 6 | 2824 | 69 | ||
| XM_006245979.2 | XP_006246041.1 | 4 - 5 | 2773 | 69 | ||
| XM_006245980.2 | XP_006246042.1 | 3 - 4 | 2749 | 69 | ||
Set Membership: |
|