Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01461292_m1
          See other DCAKD GE Assays ›
          Gene Symbol:
          Dcakd
          Gene Name:
          dephospho-CoA kinase domain containing
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 91095601 - 91126964 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01461292_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dcakd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007724.1 NP_001007725.1 4 - 5 571 78
          XM_006247532.2 XP_006247594.1 4 - 5 820 78
          XM_008768293.1 XP_008766515.1 4 - 5 618 78
          XM_008768294.1 XP_008766516.1 4 - 5 936 78
          GenBank mRNA BC079186.1 - 4 - 5 571 7131172 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003891.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dcakd

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          coenzyme A biosynthetic process
          phosphorylation
          dephospho-CoA kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P871648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422212 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P871648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P871648 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P871648 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P356855 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P871648 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82900 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16854 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747980 - -
          Affymetrix Rat Gene 1.0 ST Array 10747984 - -
          Affymetrix Rat Gene 1.0 ST Array 10747981 - -
          Affymetrix Rat Gene 1.0 ST Array 10747983 - -
          Affymetrix Rat Gene 1.0 ST Array 10747982 - -
          Affymetrix Rat Gene 1.1 ST Array 10747979 - -
          Rat Genome 230 2.0 Array 1379861_at - -
          Rat Genome 230 2.0 Array 1389274_PM_at - -
          Rat Genome 230 2.0 Array 1389274_at - -
          Rat Genome 230 2.0 Array 1379861_PM_at - -
          Affymetrix HT Rat Focus Array 1379861_PM_at - -
          Affymetrix HT Rat Focus Array 1379861_at - -
          Affymetrix HT Rat Focus Array 1389274_PM_at - -
          Affymetrix HT Rat Focus Array 1389274_at - -
          Affymetrix Rat Clariom S Assay PSR1000035181.rn.1 TC1000003891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035180.rn.1 TC1000003891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035182.rn.1 TC1000003891.rn.1 RTA TC
          Rat Expression Set 230 1389274_at - -
          Rat Expression Set 230 1389274_PM_at - -
          Rat Expression Set 230 1379861_at - -
          Rat Expression Set 230 1379861_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747979 - -
          Rat Genome U34 Set rc_AA800803_at - -
          Rat Genome U34 Set rc_AA800803_g_at - -
          Rat Genome U34 Set rc_AA900992_at - -
          Rat Genome U34 Set rc_AI102585_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22327030 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367325 - -
          RatRef-12 Expression BeadChip ILMN_67134 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline