Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01461760_m1
          See other HAUS3 GE Assays ›
          Gene Symbol:
          Haus3
          Gene Name:
          HAUS augmin-like complex, subunit 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 81837781 - 81857458 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01461760_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Haus3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109412.1 NP_001102882.1 3 - 4 1541 129
          XM_008770361.1 XP_008768583.1 2 - 3 2275 129
          XM_008770362.1 XP_008768584.1 2 - 3 1847 129
          GenBank mRNA BC162020.1 - 3 - 4 1513 9098415 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000833.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          spindle assembly
          centrosome organization

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097608 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50074 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097608 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097607 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097607 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097608 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097608 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097607 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P447814 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773557 - -
          Affymetrix Rat Gene 1.0 ST Array 10773556 - -
          Affymetrix Rat Gene 1.0 ST Array 10773558 - -
          Affymetrix Rat Gene 1.0 ST Array 10773555 - -
          Affymetrix Rat Gene 1.0 ST Array 10773554 - -
          Affymetrix Rat Gene 1.1 ST Array 10773553 - -
          Affymetrix Rat Clariom S Assay PSR1400005807.rn.1 TC1400000833.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005810.rn.1 TC1400000833.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005806.rn.1 TC1400000833.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10773553 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22363869 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline