Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01461970_m1
          See other CAR3 GE Assays ›
          Gene Symbol:
          Car3
          Gene Name:
          carbonic anhydrase 3
          Gene Aliases:
          Ca3
          Chromosome Location:
          Chr.2: 88126519 - 88136063 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01461970_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Car3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019292.4 NP_062165.2 5 - 6 514 68
          XM_006232116.1 XP_006232178.1 6 - 7 839 68
          GenBank mRNA AB030829.1 - 5 - 6 514 68
          AB030830.1 - 5 - 6 514 68
          AF037072.1 - 5 - 6 537 68
          BC061980.1 - 5 - 6 549 5597975 68
          FQ210528.1 - 5 - 6 562 68
          FQ211459.1 - 5 - 6 561 68
          FQ214846.1 - 5 - 6 559 68
          FQ214929.1 - 5 - 6 564 68
          FQ215392.1 - 4 - 5 376 68
          FQ216182.1 - 5 - 6 560 68
          FQ223846.1 - 5 - 6 560 68
          M22413.1 - 5 - 6 669 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003226.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Car3

          Gene Ontology Categories:

          Function(s) Process(es)

          one-carbon metabolic process
          response to oxidative stress
          response to ethanol
          carbonate dehydratase activity
          zinc ion binding
          nickel cation binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P244851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157740 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P244851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P244851 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P244851 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P244851 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700820751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79350 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20384 - -
          Agilent Rat cDNA Microarray (G4105A) 700820751 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822248 - -
          Affymetrix Rat Gene 1.0 ST Array 10822247 - -
          Affymetrix Rat Gene 1.0 ST Array 10822246 - -
          Affymetrix Rat Gene 1.0 ST Array 10822250 - -
          Affymetrix Rat Gene 1.0 ST Array 10822243 - -
          Affymetrix Rat Gene 1.0 ST Array 10822244 - -
          Affymetrix Rat Gene 1.0 ST Array 10822251 - -
          Affymetrix Rat Gene 1.0 ST Array 10822252 - -
          Affymetrix Rat Gene 1.0 ST Array 10822249 - -
          Affymetrix Rat Gene 1.0 ST Array 10822245 - -
          Affymetrix Rat Gene 1.1 ST Array 10822242 - -
          Rat Genome 230 2.0 Array 1386977_at - -
          Rat Genome 230 2.0 Array 1386977_PM_at - -
          Rat Genome 230 2.0 Array 1367896_PM_at - -
          Rat Genome 230 2.0 Array 1367896_at - -
          Affymetrix HT Rat Focus Array 1394551_at - -
          Affymetrix HT Rat Focus Array 1394551_PM_at - -
          Affymetrix HT Rat Focus Array 1386977_PM_at - -
          Affymetrix HT Rat Focus Array 1367896_at - -
          Affymetrix HT Rat Focus Array 1367896_PM_at - -
          Affymetrix HT Rat Focus Array 1386977_at - -
          Affymetrix Rat Clariom S Assay PSR0200022265.rn.1 TC0200003226.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022264.rn.1 TC0200003226.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022267.rn.1 TC0200003226.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022261.rn.1 TC0200003226.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022263.rn.1 TC0200003226.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022266.rn.1 TC0200003226.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022262.rn.1 TC0200003226.rn.1 RTA TC
          Rat Expression Set 230 1386977_PM_at - -
          Rat Expression Set 230 1386977_at - -
          Rat Expression Set 230 1367896_at - -
          Rat Expression Set 230 1367896_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822242 - -
          Rat Genome U34 Set AF037072_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20871438 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368331 - -
          RatRef-12 Expression BeadChip ILMN_49903 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline