Search
Search

RGD1307100| Gene Symbol: RGD1307100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037184.1 | NP_001032261.1 | 2 - 3 | 625 | 102 | |
| XM_006232275.2 | XP_006232337.1 | 86 - 87 | 15085 | 102 | ||
| XM_008760914.1 | XP_008759136.1 | 86 - 87 | 15085 | 102 | ||
| XM_008760915.1 | XP_008759137.1 | 86 - 87 | 15082 | 102 | ||
| XM_008760916.1 | XP_008759138.1 | 85 - 86 | 15034 | 102 | ||
| XM_008760917.1 | XP_008759139.1 | 85 - 86 | 15022 | 102 | ||
| XM_008760918.1 | XP_008759140.1 | 85 - 86 | 14947 | 102 | ||
| XM_008760919.1 | XP_008759141.1 | 85 - 86 | 14883 | 102 | ||
| GenBank mRNA | BC100102.1 | - | 2 - 3 | 625 | 7377390 | 102 |
Set Membership: |
|