Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462275_m1
          See other UTP14A GE Assays ›
          Gene Symbol:
          Utp14a
          Gene Name:
          UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 135092123 - 135117409 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462275_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Utp14a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014113.2 NP_001014135.2 14 - 15 2067 119
          GenBank mRNA BC088326.1 - 4 - 5 1227 7365084 119
          BC099799.1 - 14 - 15 2067 7936638 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001144.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Utp14a

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          maturation of SSU-rRNA
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P076357 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139162 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014165 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076357 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014165 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076357 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014165 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076357 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P192382 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P183039 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13162 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94919 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935335 - -
          Affymetrix Rat Gene 1.0 ST Array 10935333 - -
          Affymetrix Rat Gene 1.0 ST Array 10935327 - -
          Affymetrix Rat Gene 1.0 ST Array 10935324 - -
          Affymetrix Rat Gene 1.0 ST Array 10935332 - -
          Affymetrix Rat Gene 1.0 ST Array 10935330 - -
          Affymetrix Rat Gene 1.0 ST Array 10935337 - -
          Affymetrix Rat Gene 1.0 ST Array 10935331 - -
          Affymetrix Rat Gene 1.0 ST Array 10935334 - -
          Affymetrix Rat Gene 1.0 ST Array 10935328 - -
          Affymetrix Rat Gene 1.0 ST Array 10935326 - -
          Affymetrix Rat Gene 1.0 ST Array 10935325 - -
          Affymetrix Rat Gene 1.0 ST Array 10935329 - -
          Affymetrix Rat Gene 1.0 ST Array 10935336 - -
          Affymetrix Rat Gene 1.1 ST Array 10935323 - -
          Rat Genome 230 2.0 Array 1383250_PM_at - -
          Rat Genome 230 2.0 Array 1383250_at - -
          Affymetrix HT Rat Focus Array 1383250_at - -
          Affymetrix HT Rat Focus Array 1383250_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00007020.rn.1 TC0X00001144.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007022.rn.1 TC0X00001144.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007021.rn.1 TC0X00001144.rn.1 RTA TC
          Rat Expression Set 230 1383250_at - -
          Rat Expression Set 230 1383250_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10935323 - -
          Rat Genome U34 Set rc_AI045862_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22307722 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371924 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370255 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline