Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462455_m1
          See other MILL1 GE Assays ›
          Gene Symbol:
          Mill1
          Gene Name:
          MHC I like leukocyte 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 79631273 - 79657943 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462455_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mill1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011931.2 NP_001011931.1 2 - 3 216 89
          XM_008758906.1 XP_008757128.1 3 - 4 279 89
          XM_008758907.1 XP_008757129.1 2 - 3 302 89
          GenBank mRNA AB113960.1 - 2 - 3 225 89
          AB113962.1 - 2 - 3 216 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000965.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mill1

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          immune response-activating cell surface receptor signaling pathway
          antigen processing and presentation of peptide antigen via MHC class I
          immune response
          response to oxidative stress
          response to heat
          antigen processing and presentation
          response to retinoic acid
          gamma-delta T cell activation
          negative regulation of defense response to virus by host
          antigen binding
          peptide antigen binding
          natural killer cell lectin-like receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P389809 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P389810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163364 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P389809 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P389809 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P389809 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468540 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25192 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704645 - -
          Affymetrix Rat Gene 1.0 ST Array 10704644 - -
          Affymetrix Rat Gene 1.0 ST Array 10704648 - -
          Affymetrix Rat Gene 1.0 ST Array 10704643 - -
          Affymetrix Rat Gene 1.0 ST Array 10704647 - -
          Affymetrix Rat Gene 1.0 ST Array 10704646 - -
          Affymetrix Rat Gene 1.0 ST Array 10704649 - -
          Affymetrix Rat Gene 1.1 ST Array 10704642 - -
          Affymetrix Rat Clariom S Assay PSR0100006708.rn.1 TC0100000965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006707.rn.1 TC0100000965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006709.rn.1 TC0100000965.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10704642 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375280 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376883 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline