Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462646_m1
          See other CLEC4A3 GE Assays ›
          Gene Symbol:
          Clec4a3
          Gene Name:
          C-type lectin domain family 4, member A3
          Gene Aliases:
          Dcir3
          Chromosome Location:
          Chr.4: 155913005 - 155923288 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462646_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec4a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005891.1 NP_001005891.1 4 - 5 488 104
          XM_006237321.2 XP_006237383.1 4 - 5 697 104
          GenBank mRNA AY397671.1 - 4 - 5 488 104
          AY494062.1 - 4 - 5 489 104
          FQ230051.1 - 4 - 5 583 104
          FQ234061.1 - 4 - 5 613 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003707.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec4a3

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P101301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42264 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101301 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P101301 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104632 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858560 - -
          Affymetrix Rat Gene 1.0 ST Array 10858565 - -
          Affymetrix Rat Gene 1.0 ST Array 10858561 - -
          Affymetrix Rat Gene 1.0 ST Array 10858564 - -
          Affymetrix Rat Gene 1.0 ST Array 10858563 - -
          Affymetrix Rat Gene 1.0 ST Array 10858562 - -
          Affymetrix Rat Gene 1.1 ST Array 10858559 - -
          Rat Genome 230 2.0 Array 1389553_at - -
          Rat Genome 230 2.0 Array 1389553_PM_at - -
          Affymetrix HT Rat Focus Array 1389553_PM_at - -
          Affymetrix HT Rat Focus Array 1389553_at - -
          Affymetrix Rat Clariom S Assay PSR0400026164.rn.1 TC0400003707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026165.rn.1 TC0400003707.rn.1 RTA TC
          Rat Expression Set 230 1389553_at - -
          Rat Expression Set 230 1389553_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858559 - -
          Rat Genome U34 Set rc_AI230142_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21115911 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376838 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline