Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01463046_m1
          See other AMN1 GE Assays ›
          Gene Symbol:
          Amn1
          Gene Name:
          antagonist of mitotic exit network 1 homolog (S. cerevisiae)
          Gene Aliases:
          RGD1308119
          Chromosome Location:
          Chr.4: 183670589 - 183697531 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01463046_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008333.2 NP_001008334.2 5 - 6 636 106
          XM_006237700.1 XP_006237762.1 5 - 6 1122 106
          GenBank mRNA BC086357.1 - 5 - 6 606 7303722 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004138.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Amn1

          Panther Classification:

          Molecular Function -

          ATP synthase primary active transporter transporter

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P382363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5004 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P382363 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P382363 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P382363 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P382363 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22811 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96945 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59123 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867013 - -
          Affymetrix Rat Gene 1.0 ST Array 10867017 - -
          Affymetrix Rat Gene 1.0 ST Array 10867014 - -
          Affymetrix Rat Gene 1.0 ST Array 10867011 - -
          Affymetrix Rat Gene 1.0 ST Array 10867012 - -
          Affymetrix Rat Gene 1.0 ST Array 10867015 - -
          Affymetrix Rat Gene 1.0 ST Array 10867016 - -
          Affymetrix Rat Gene 1.1 ST Array 10867010 - -
          Rat Genome 230 2.0 Array 1376714_at - -
          Rat Genome 230 2.0 Array 1381961_PM_at - -
          Rat Genome 230 2.0 Array 1381961_at - -
          Rat Genome 230 2.0 Array 1376714_PM_at - -
          Rat Genome 230 2.0 Array 1380192_PM_at - -
          Rat Genome 230 2.0 Array 1380192_at - -
          Affymetrix HT Rat Focus Array 1376714_PM_at - -
          Affymetrix HT Rat Focus Array 1376714_at - -
          Affymetrix HT Rat Focus Array 1380192_PM_at - -
          Affymetrix HT Rat Focus Array 1380192_at - -
          Affymetrix Rat Clariom S Assay PSR0400029394.rn.1 TC0400004138.rn.1 RTA TC
          Rat Expression Set 230 1376714_at - -
          Rat Expression Set 230 1381961_PM_at - -
          Rat Expression Set 230 1376714_PM_at - -
          Rat Expression Set 230 1381961_at - -
          Rat Expression Set 230 1380192_PM_at - -
          Rat Expression Set 230 1380192_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867010 - -
          Rat Genome U34 Set rc_AI236072_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21811580 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357643 - -
          RatRef-12 Expression BeadChip ILMN_70160 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline