Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01463509_m1
          See other MFAP5 GE Assays ›
          Gene Symbol:
          Mfap5
          Gene Name:
          microfibrillar associated protein 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 155313671 - 155336228 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01463509_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfap5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108644.1 NP_001102114.1 10 - 11 537 82
          GenBank mRNA FQ229611.1 - 9 - 10 643 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001725.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          extracellular fibril organization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133769 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14973 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14973 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14973 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858504 - -
          Affymetrix Rat Gene 1.0 ST Array 10858508 - -
          Affymetrix Rat Gene 1.0 ST Array 10858500 - -
          Affymetrix Rat Gene 1.0 ST Array 10858505 - -
          Affymetrix Rat Gene 1.0 ST Array 10858507 - -
          Affymetrix Rat Gene 1.0 ST Array 10858502 - -
          Affymetrix Rat Gene 1.0 ST Array 10858503 - -
          Affymetrix Rat Gene 1.0 ST Array 10858501 - -
          Affymetrix Rat Gene 1.0 ST Array 10858506 - -
          Affymetrix Rat Gene 1.1 ST Array 10858499 - -
          Rat Genome 230 2.0 Array 1373674_PM_at - -
          Rat Genome 230 2.0 Array 1373674_at - -
          Affymetrix HT Rat Focus Array 1373674_PM_at - -
          Affymetrix HT Rat Focus Array 1373674_at - -
          Affymetrix Rat Clariom S Assay PSR0400012534.rn.1 TC0400001725.rn.1 RTA TC
          Rat Expression Set 230 1373674_PM_at - -
          Rat Expression Set 230 1373674_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858499 - -
          Rat Genome U34 Set rc_AI235874_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21297493 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365298 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline