Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01463598_m1
          See other NBR1 GE Assays ›
          Gene Symbol:
          Nbr1
          Gene Name:
          neighbor of Brca1 gene 1
          Gene Aliases:
          Ca125, RGD1311421
          Chromosome Location:
          Chr.10: 89455024 - 89483791 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01463598_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nbr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024765.1 NP_001019936.1 18 - 19 2280 132
          XM_006247321.2 XP_006247383.1 18 - 19 2562 132
          XM_006247322.2 XP_006247384.1 18 - 19 2477 132
          XM_006247323.2 XP_006247385.1 18 - 19 2334 132
          XM_006247324.2 XP_006247386.1 18 - 19 2281 132
          XM_006247325.2 XP_006247387.1 17 - 18 2326 132
          XM_006247326.2 XP_006247388.1 18 - 19 2432 132
          GenBank mRNA BC095903.1 - 18 - 19 2280 7096697 132
          FQ211883.1 - 18 - 19 2280 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004375.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nbr1

          Gene Ontology Categories:

          Function(s) Process(es)

          macroautophagy
          regulation of bone mineralization
          regulation of stress-activated MAPK cascade
          negative regulation of osteoblast differentiation
          protein oligomerization
          zinc ion binding
          ubiquitin binding
          mitogen-activated protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137148 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006798 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006798 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006798 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P471440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36442 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10738436 - -
          Affymetrix Rat Gene 1.0 ST Array 10738447 - -
          Affymetrix Rat Gene 1.0 ST Array 10738442 - -
          Affymetrix Rat Gene 1.0 ST Array 10738441 - -
          Affymetrix Rat Gene 1.0 ST Array 10738445 - -
          Affymetrix Rat Gene 1.0 ST Array 10738448 - -
          Affymetrix Rat Gene 1.0 ST Array 10738438 - -
          Affymetrix Rat Gene 1.0 ST Array 10738444 - -
          Affymetrix Rat Gene 1.0 ST Array 10738433 - -
          Affymetrix Rat Gene 1.0 ST Array 10738437 - -
          Affymetrix Rat Gene 1.0 ST Array 10738434 - -
          Affymetrix Rat Gene 1.0 ST Array 10738440 - -
          Affymetrix Rat Gene 1.0 ST Array 10738450 - -
          Affymetrix Rat Gene 1.0 ST Array 10738439 - -
          Affymetrix Rat Gene 1.0 ST Array 10738432 - -
          Affymetrix Rat Gene 1.0 ST Array 10738446 - -
          Affymetrix Rat Gene 1.0 ST Array 10772798 - -
          Affymetrix Rat Gene 1.0 ST Array 10738443 - -
          Affymetrix Rat Gene 1.0 ST Array 10738431 - -
          Affymetrix Rat Gene 1.0 ST Array 10738435 - -
          Affymetrix Rat Gene 1.0 ST Array 10738449 - -
          Affymetrix Rat Gene 1.1 ST Array 10738430 - -
          Affymetrix Rat Gene 1.1 ST Array 10772797 - -
          Rat Genome 230 2.0 Array 1372497_PM_at - -
          Rat Genome 230 2.0 Array 1381509_PM_at - -
          Rat Genome 230 2.0 Array 1381509_at - -
          Rat Genome 230 2.0 Array 1372497_at - -
          Affymetrix HT Rat Focus Array 1381509_at - -
          Affymetrix HT Rat Focus Array 1372497_at - -
          Affymetrix HT Rat Focus Array 1372497_PM_at - -
          Affymetrix HT Rat Focus Array 1381509_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000040770.rn.1 TC1000004375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040771.rn.1 TC1000004375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040769.rn.1 TC1000004375.rn.1 RTA TC
          Rat Expression Set 230 1372497_at - -
          Rat Expression Set 230 1372497_PM_at - -
          Rat Expression Set 230 1381509_at - -
          Rat Expression Set 230 1381509_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10738430 - -
          Rat Genome U34 Set rc_AA956140_at - -
          Rat Genome U34 Set rc_AA925045_g_at - -
          Rat Genome U34 Set rc_AA925045_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21140070 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline