Search
Search
| Gene Symbol: Psmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008281.1 | NP_001008282.1 | 6 - 7 | 1109 | 64 | |
| XM_006247272.1 | XP_006247334.1 | 6 - 7 | 1324 | 64 | ||
| GenBank mRNA | BC085881.1 | - | 6 - 7 | 1109 | 7191926 | 64 |
Set Membership: |
|