Search
Search
| Gene Symbol: Cdr2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247771.2 | XP_006247833.1 | 3 - 4 | 796 | 66 | |
| XM_006247772.1 | XP_006247834.1 | 3 - 4 | 695 | 66 | ||
| XM_008768410.1 | XP_008766632.1 | 3 - 4 | 515 | 66 | ||
| XM_340933.7 | XP_340934.5 | 3 - 4 | 797 | 66 | ||
Set Membership: |
|