Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01464149_g1
          See other CLEC10A GE Assays ›
          Gene Symbol:
          Clec10a
          Gene Name:
          C-type lectin domain family 10, member A
          Gene Aliases:
          Mgl, Mgl1, Mgll
          Chromosome Location:
          Chr.10: 56764892 - 56769447 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01464149_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec10a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022393.2 NP_071788.2 9 - 10 1078 110
          XM_006246803.2 XP_006246865.1 9 - 10 1116 110
          XM_008767867.1 XP_008766089.1 9 - 10 1116 110
          XM_008767868.1 XP_008766090.1 9 - 10 960 110
          XM_008767869.1 XP_008766091.1 9 - 10 957 110
          GenBank mRNA BC091151.1 - 9 - 10 1078 7375234 110
          FQ213042.1 - 9 - 10 950 110
          FQ228437.1 - 9 - 10 936 110
          J05495.1 - 9 - 10 938 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000952.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec10a

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          connective tissue replacement involved in inflammatory response wound healing
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P520349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P520349 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P520349 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P520349 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P520349 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700525863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41206 - -
          Agilent Rat cDNA Microarray (G4105A) 700525863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735192 - -
          Affymetrix Rat Gene 1.0 ST Array 10735185 - -
          Affymetrix Rat Gene 1.0 ST Array 10735187 - -
          Affymetrix Rat Gene 1.0 ST Array 10735184 - -
          Affymetrix Rat Gene 1.0 ST Array 10735189 - -
          Affymetrix Rat Gene 1.0 ST Array 10735183 - -
          Affymetrix Rat Gene 1.0 ST Array 10735194 - -
          Affymetrix Rat Gene 1.0 ST Array 10735191 - -
          Affymetrix Rat Gene 1.0 ST Array 10735186 - -
          Affymetrix Rat Gene 1.0 ST Array 10735188 - -
          Affymetrix Rat Gene 1.0 ST Array 10735190 - -
          Affymetrix Rat Gene 1.0 ST Array 10735193 - -
          Affymetrix Rat Gene 1.1 ST Array 10735182 - -
          Rat Genome 230 2.0 Array 1368464_PM_at - -
          Rat Genome 230 2.0 Array 1368464_at - -
          Affymetrix HT Rat Focus Array 1368464_at - -
          Affymetrix HT Rat Focus Array 1368464_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000008438.rn.1 TC1000000952.rn.1 RTA TC
          Rat Expression Set 230 1368464_at - -
          Rat Expression Set 230 1368464_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735182 - -
          Rat Genome U34 Set J05495_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21979497 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374266 - -
          RatRef-12 Expression BeadChip ILMN_59086 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline