Search
Search
| Gene Symbol: Eme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750789.3 | XP_003750837.1 | 2 - 3 | 433 | 146 | |
| XM_008767602.1 | XP_008765824.1 | 2 - 3 | 433 | 146 | ||
| XM_008767603.1 | XP_008765825.1 | 2 - 3 | 436 | 146 | ||
| XM_008767604.1 | XP_008765826.1 | 2 - 3 | 436 | 146 | ||
| XM_008767605.1 | XP_008765827.1 | 2 - 3 | 399 | 146 | ||
| GenBank mRNA | BC158841.1 | - | 2 - 3 | 379 | 146 | |
Set Membership: |
|