Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01464595_m1
          See other SERPINF2 GE Assays ›
          Gene Symbol:
          Serpinf2
          Gene Name:
          serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 62264247 - 62272476 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01464595_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011892.1 NP_001011892.1 8 - 9 900 79
          XM_006246675.1 XP_006246737.1 8 - 9 1045 79
          GenBank mRNA BC079362.1 - 8 - 9 900 7109243 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003244.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinf2

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of blood vessel size by renin-angiotensin
          response to organic substance
          negative regulation of plasminogen activation
          negative regulation of endopeptidase activity
          collagen fibril organization
          positive regulation of collagen biosynthetic process
          positive regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          blood vessel morphogenesis
          positive regulation of smooth muscle cell proliferation
          positive regulation of stress fiber assembly
          negative regulation of fibrinolysis
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of transforming growth factor beta production
          protease binding
          serine-type endopeptidase inhibitor activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P762292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194067 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P762292 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P762292 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P762292 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P762292 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P176025 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17841 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744951 - -
          Affymetrix Rat Gene 1.0 ST Array 10744952 - -
          Affymetrix Rat Gene 1.0 ST Array 10744957 - -
          Affymetrix Rat Gene 1.0 ST Array 10744950 - -
          Affymetrix Rat Gene 1.0 ST Array 10744948 - -
          Affymetrix Rat Gene 1.0 ST Array 10744956 - -
          Affymetrix Rat Gene 1.0 ST Array 10744954 - -
          Affymetrix Rat Gene 1.0 ST Array 10744953 - -
          Affymetrix Rat Gene 1.0 ST Array 10744955 - -
          Affymetrix Rat Gene 1.0 ST Array 10744949 - -
          Affymetrix Rat Gene 1.1 ST Array 10744947 - -
          Rat Genome 230 2.0 Array 1377033_PM_at - -
          Rat Genome 230 2.0 Array 1377033_at - -
          Affymetrix HT Rat Focus Array 1377033_at - -
          Affymetrix HT Rat Focus Array 1377033_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000029123.rn.1 TC1000003244.rn.1 RTA TC
          Rat Expression Set 230 1377033_PM_at - -
          Rat Expression Set 230 1377033_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744947 - -
          Rat Genome U34 Set rc_AI233031_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21490913 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376805 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375995 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline