Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01464841_m1
          See other POLG2 GE Assays ›
          Gene Symbol:
          Polg2
          Gene Name:
          polymerase (DNA directed), gamma 2, accessory subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 94968836 - 94979259 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01464841_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107060.1 NP_001100530.1 1 - 2 876 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003965.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          aminoacyl-tRNA synthetase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial DNA replication
          DNA biosynthetic process
          DNA-directed DNA polymerase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P517554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P517554 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P517554 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P517554 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P517554 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748344 - -
          Affymetrix Rat Gene 1.0 ST Array 10748342 - -
          Affymetrix Rat Gene 1.0 ST Array 10748339 - -
          Affymetrix Rat Gene 1.0 ST Array 10748341 - -
          Affymetrix Rat Gene 1.0 ST Array 10748345 - -
          Affymetrix Rat Gene 1.0 ST Array 10748346 - -
          Affymetrix Rat Gene 1.0 ST Array 10748340 - -
          Affymetrix Rat Gene 1.0 ST Array 10748343 - -
          Affymetrix Rat Gene 1.1 ST Array 10748338 - -
          Rat Genome 230 2.0 Array 1375504_at - -
          Rat Genome 230 2.0 Array 1375504_PM_at - -
          Affymetrix HT Rat Focus Array 1375504_at - -
          Affymetrix HT Rat Focus Array 1375504_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000036079.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036076.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036075.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036084.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036078.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036086.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036073.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036082.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036087.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036083.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036074.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036077.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036085.rn.1 TC1000003965.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000036080.rn.1 TC1000003965.rn.1 RTA TC
          Rat Expression Set 230 1375504_PM_at - -
          Rat Expression Set 230 1375504_at - -
          Affymetrix Rat Gene 1.0 ST Array 10748338 - -
          Rat Genome U34 Set rc_AA892950_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline