Search
Search
| Gene Symbol: Nprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034936.1 | NP_001030108.1 | 6 - 7 | 719 | 62 | |
| XM_006246008.2 | XP_006246070.1 | 7 - 8 | 841 | 62 | ||
| XM_006246009.2 | XP_006246071.1 | 7 - 8 | 841 | 62 | ||
| XM_006246010.2 | XP_006246072.1 | 6 - 7 | 801 | 62 | ||
| XM_006246011.2 | XP_006246073.1 | 7 - 8 | 653 | 62 | ||
| XM_006246012.2 | XP_006246074.1 | 3 - 4 | 1834 | 62 | ||
| XM_006246013.2 | XP_006246075.1 | 5 - 6 | 2283 | 62 | ||
| XM_006246014.2 | XP_006246076.1 | 5 - 6 | 2184 | 62 | ||
| XM_006246015.2 | XP_006246077.1 | 5 - 6 | 2485 | 62 | ||
| XM_006246016.2 | XP_006246078.1 | 7 - 8 | 853 | 62 | ||
| XM_006246017.2 | XP_006246079.1 | 4 - 5 | 375 | 62 | ||
| GenBank mRNA | BC099803.1 | - | 6 - 7 | 719 | 7937864 | 62 |
Set Membership: |
|