Search
Search
| Gene Symbol: Zfp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012008.1 | NP_001012008.1 | 3 - 4 | 708 | 81 | |
| XM_006246695.2 | XP_006246757.1 | 4 - 5 | 878 | 81 | ||
| XM_006246696.2 | XP_006246758.1 | 4 - 5 | 878 | 81 | ||
| XM_006246697.2 | XP_006246759.1 | 4 - 5 | 878 | 81 | ||
| GenBank mRNA | BC081874.1 | - | 3 - 4 | 708 | 7107522 | 81 |
Set Membership: |
|