Search
Search
| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770921.1 | XP_008769143.1 | 565 | 61 | ||
| XM_008770922.1 | XP_008769144.1 | 785 | 61 | |||
| XM_008770923.1 | XP_008769145.1 | 563 | 61 | |||
| XM_008770924.1 | XP_008769146.1 | 715 | 61 | |||
Set Membership: |
|