Search
Search
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005762.1 | NP_001005762.1 | 18 - 19 | 1774 | 67 | |
| XM_006252241.2 | XP_006252303.1 | 19 - 20 | 3458 | 67 | ||
| XM_006252242.1 | XP_006252304.1 | 18 - 19 | 1865 | 67 | ||
| XM_006252244.2 | XP_006252306.1 | 17 - 18 | 2093 | 67 | ||
| XM_006252246.1 | XP_006252308.1 | 18 - 19 | 1810 | 67 | ||
| XM_006252248.1 | XP_006252310.1 | 6 - 7 | 658 | 67 | ||
| XM_008770819.1 | XP_008769041.1 | 16 - 17 | 3567 | 67 | ||
| GenBank mRNA | BC083666.1 | - | 18 - 19 | 1774 | 7135547 | 67 |
Set Membership: |
|