Search
Search
| Gene Symbol: Abcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133411.1 | NP_596902.1 | 7 - 8 | 914 | 64 | |
| XM_008770939.1 | XP_008769161.1 | 7 - 8 | 1302 | 64 | ||
| GenBank mRNA | AF534126.1 | - | 7 - 8 | 925 | 64 | |
| AY533524.1 | - | 7 - 8 | 914 | 64 | ||
| AY533525.1 | - | 6 - 7 | 803 | 64 | ||
Set Membership: |
|