Search
Search
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053766.1 | NP_446218.1 | 6 - 7 | 732 | 74 | |
| XM_008770840.1 | XP_008769062.1 | 3 - 4 | 1248 | 74 | ||
| XM_008770841.1 | XP_008769063.1 | 4 - 5 | 751 | 74 | ||
| GenBank mRNA | BC089052.1 | - | 6 - 7 | 732 | 7307299 | 74 |
Set Membership: |
|