Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466151_m1
          See other NUFIP1 GE Assays ›
          Gene Symbol:
          Nufip1
          Gene Name:
          nuclear fragile X mental retardation protein interacting protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 58171163 - 58201888 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01466151_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nufip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007758.1 NP_001007759.1 9 - 10 1346 131
          GenBank mRNA BC082111.1 - 9 - 10 1346 7135272 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000857.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nufip1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          box C/D snoRNP assembly
          positive regulation of transcription from RNA polymerase II promoter
          RNA binding
          snoRNA binding
          protein binding, bridging
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P407707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176106 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146848 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P407707 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P407707 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P407707 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P407707 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P695017 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68900 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28768 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781578 - -
          Affymetrix Rat Gene 1.0 ST Array 10781572 - -
          Affymetrix Rat Gene 1.0 ST Array 10781571 - -
          Affymetrix Rat Gene 1.0 ST Array 10781575 - -
          Affymetrix Rat Gene 1.0 ST Array 10781574 - -
          Affymetrix Rat Gene 1.0 ST Array 10781570 - -
          Affymetrix Rat Gene 1.0 ST Array 10781573 - -
          Affymetrix Rat Gene 1.0 ST Array 10781576 - -
          Affymetrix Rat Gene 1.0 ST Array 10781569 - -
          Affymetrix Rat Gene 1.0 ST Array 10781577 - -
          Affymetrix Rat Gene 1.1 ST Array 10781568 - -
          Rat Genome 230 2.0 Array 1376250_PM_at - -
          Rat Genome 230 2.0 Array 1394371_at - -
          Rat Genome 230 2.0 Array 1394371_PM_at - -
          Rat Genome 230 2.0 Array 1376250_at - -
          Affymetrix HT Rat Focus Array 1394371_at - -
          Affymetrix HT Rat Focus Array 1376250_at - -
          Affymetrix HT Rat Focus Array 1394371_PM_at - -
          Affymetrix HT Rat Focus Array 1376250_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500005937.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005931.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005921.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005935.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005923.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005926.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005928.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005925.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005929.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005930.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005936.rn.1 TC1500000857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005922.rn.1 TC1500000857.rn.1 RTA TC
          Rat Expression Set 230 1394371_at - -
          Rat Expression Set 230 1376250_PM_at - -
          Rat Expression Set 230 1376250_at - -
          Rat Expression Set 230 1394371_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781568 - -
          Rat Genome U34 Set rc_AI103747_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21574332 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358584 - -
          RatRef-12 Expression BeadChip ILMN_55470 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline