Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466456_m1
          See other MED13 GE Assays ›
          Gene Symbol:
          Med13
          Gene Name:
          mediator complex subunit 13
          Gene Aliases:
          Thrap1
          Chromosome Location:
          Chr.10: 73698789 - 73787084 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01466456_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Med13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107035.1 NP_001100505.1 19 - 20 4773 72
          XM_006247066.2 XP_006247128.1 19 - 20 4777 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003468.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription initiation from RNA polymerase II promoter
          biological_process
          intracellular steroid hormone receptor signaling pathway
          androgen receptor signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II transcription cofactor activity
          molecular_function
          transcription cofactor activity
          transcription coactivator activity
          receptor activity
          thyroid hormone receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P445972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137981 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177341 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P445972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P445972 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P445972 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P445972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70497 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745909 - -
          Affymetrix Rat Gene 1.0 ST Array 10745908 - -
          Affymetrix Rat Gene 1.0 ST Array 10745905 - -
          Affymetrix Rat Gene 1.0 ST Array 10745921 - -
          Affymetrix Rat Gene 1.0 ST Array 10745918 - -
          Affymetrix Rat Gene 1.0 ST Array 10745912 - -
          Affymetrix Rat Gene 1.0 ST Array 10745914 - -
          Affymetrix Rat Gene 1.0 ST Array 10745899 - -
          Affymetrix Rat Gene 1.0 ST Array 10745927 - -
          Affymetrix Rat Gene 1.0 ST Array 10745902 - -
          Affymetrix Rat Gene 1.0 ST Array 10745911 - -
          Affymetrix Rat Gene 1.0 ST Array 10745904 - -
          Affymetrix Rat Gene 1.0 ST Array 10745926 - -
          Affymetrix Rat Gene 1.0 ST Array 10745906 - -
          Affymetrix Rat Gene 1.0 ST Array 10745913 - -
          Affymetrix Rat Gene 1.0 ST Array 10745922 - -
          Affymetrix Rat Gene 1.0 ST Array 10745903 - -
          Affymetrix Rat Gene 1.0 ST Array 10745916 - -
          Affymetrix Rat Gene 1.0 ST Array 10745900 - -
          Affymetrix Rat Gene 1.0 ST Array 10745907 - -
          Affymetrix Rat Gene 1.0 ST Array 10745915 - -
          Affymetrix Rat Gene 1.0 ST Array 10745924 - -
          Affymetrix Rat Gene 1.0 ST Array 10745917 - -
          Affymetrix Rat Gene 1.0 ST Array 10745901 - -
          Affymetrix Rat Gene 1.0 ST Array 10745923 - -
          Affymetrix Rat Gene 1.0 ST Array 10745910 - -
          Affymetrix Rat Gene 1.0 ST Array 10745925 - -
          Affymetrix Rat Gene 1.0 ST Array 10745919 - -
          Affymetrix Rat Gene 1.0 ST Array 10745920 - -
          Affymetrix Rat Gene 1.1 ST Array 10745897 - -
          Rat Genome 230 2.0 Array 1386156_PM_at - -
          Rat Genome 230 2.0 Array 1379567_PM_at - -
          Rat Genome 230 2.0 Array 1385091_PM_at - -
          Rat Genome 230 2.0 Array 1379567_at - -
          Rat Genome 230 2.0 Array 1386156_at - -
          Rat Genome 230 2.0 Array 1385091_at - -
          Affymetrix HT Rat Focus Array 1379567_at - -
          Affymetrix HT Rat Focus Array 1385091_at - -
          Affymetrix HT Rat Focus Array 1385091_PM_at - -
          Affymetrix HT Rat Focus Array 1379567_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000031025.rn.1 TC1000003468.rn.1 RTA TC
          Rat Expression Set 230 1386156_PM_at - -
          Rat Expression Set 230 1386156_at - -
          Rat Expression Set 230 1385091_PM_at - -
          Rat Expression Set 230 1379567_at - -
          Rat Expression Set 230 1385091_at - -
          Rat Expression Set 230 1379567_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745897 - -
          Rat Genome U34 Set rc_AI639451_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20911918 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372402 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline