Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466623_gH
          See other SMARCE1 GE Assays ›
          Gene Symbol:
          Smarce1
          Gene Name:
          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1
          Gene Aliases:
          BAF57, RGD1304726
          Chromosome Location:
          Chr.10: 87116827 - 87138248 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01466623_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smarce1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024993.1 NP_001020164.1 6 - 7 534 82
          XM_008768051.1 XP_008766273.1 6 - 7 876 82
          GenBank mRNA BC091314.1 - 6 - 7 590 7312354 82
          BC092210.1 - 6 - 7 534 7311688 82
          FQ213000.1 - 6 - 7 522 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smarce1

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome disassembly
          chromatin remodeling
          regulation of transcription, DNA-templated
          spermatid development
          brain development
          metabolic process
          ATP-dependent chromatin remodeling
          negative regulation of transcription, DNA-templated
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          DNA binding
          chromatin binding
          RNA binding
          N-acetyltransferase activity
          ligand-dependent nuclear receptor binding
          nucleosomal DNA binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157144 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56392 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157144 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157149 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157149 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157144 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157149 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157144 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P755445 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 866 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747034 - -
          Affymetrix Rat Gene 1.0 ST Array 10747030 - -
          Affymetrix Rat Gene 1.0 ST Array 10747031 - -
          Affymetrix Rat Gene 1.0 ST Array 10747032 - -
          Affymetrix Rat Gene 1.0 ST Array 10747033 - -
          Affymetrix Rat Gene 1.0 ST Array 10810408 - -
          Affymetrix Rat Gene 1.1 ST Array 10747029 - -
          Affymetrix Rat Gene 1.1 ST Array 10810407 - -
          Rat Genome 230 2.0 Array 1378203_at - -
          Rat Genome 230 2.0 Array 1378203_PM_at - -
          Rat Genome 230 2.0 Array 1372938_at - -
          Rat Genome 230 2.0 Array 1383806_PM_at - -
          Rat Genome 230 2.0 Array 1372938_PM_at - -
          Rat Genome 230 2.0 Array 1383806_at - -
          Affymetrix HT Rat Focus Array 1383806_PM_at - -
          Affymetrix HT Rat Focus Array 1383806_at - -
          Affymetrix HT Rat Focus Array 1372938_PM_at - -
          Affymetrix HT Rat Focus Array 1378203_PM_at - -
          Affymetrix HT Rat Focus Array 1372938_at - -
          Affymetrix HT Rat Focus Array 1378203_at - -
          Affymetrix Rat Clariom S Assay PSR1000033430.rn.1 TC1000003732.rn.1 RTA TC
          Rat Expression Set 230 1372938_PM_at - -
          Rat Expression Set 230 1372938_at - -
          Rat Expression Set 230 1383806_PM_at - -
          Rat Expression Set 230 1383806_at - -
          Rat Expression Set 230 1378203_PM_at - -
          Rat Expression Set 230 1378203_at - -
          Affymetrix Rat Gene 1.0 ST Array 10810407 - -
          Affymetrix Rat Gene 1.0 ST Array 10747029 - -
          Rat Genome U34 Set rc_AI010410_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20694107 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355392 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline