Search
Search
| Gene Symbol: Rad51c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129777.1 | NP_001123249.1 | 1 - 2 | 141 | 77 | |
| XM_006247106.1 | XP_006247168.1 | 1 - 2 | 219 | 77 | ||
| GenBank mRNA | BC166862.1 | - | 1 - 2 | 48 | 77 | |
Set Membership: |
|