Search
Search
| Gene Symbol: Lrrc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008280.1 | NP_001008281.1 | 2 - 3 | 377 | 97 | |
| GenBank mRNA | BC086530.1 | - | 2 - 3 | 377 | 7304726 | 97 |
| D13623.1 | - | 2 - 3 | 414 | 97 | ||
Set Membership: |
|