Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466809_m1
          See other PELP1 GE Assays ›
          Gene Symbol:
          Pelp1
          Gene Name:
          proline, glutamate and leucine rich protein 1
          Gene Aliases:
          RGD1306320
          Chromosome Location:
          Chr.10: 56988076 - 57005111 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01466809_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pelp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024270.2 NP_001019441.2 5 - 6 650 61
          GenBank mRNA AY970831.2 - 5 - 6 648 61
          BC101890.1 - 5 - 6 650 7930679 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003127.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pelp1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cellular response to estrogen stimulus
          chromatin binding
          transcription factor binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P286135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P286135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P286135 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P286135 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P396507 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37707 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94321 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744447 - -
          Affymetrix Rat Gene 1.0 ST Array 10744458 - -
          Affymetrix Rat Gene 1.0 ST Array 10744449 - -
          Affymetrix Rat Gene 1.0 ST Array 10744454 - -
          Affymetrix Rat Gene 1.0 ST Array 10744442 - -
          Affymetrix Rat Gene 1.0 ST Array 10744459 - -
          Affymetrix Rat Gene 1.0 ST Array 10744444 - -
          Affymetrix Rat Gene 1.0 ST Array 10744443 - -
          Affymetrix Rat Gene 1.0 ST Array 10744457 - -
          Affymetrix Rat Gene 1.0 ST Array 10744456 - -
          Affymetrix Rat Gene 1.0 ST Array 10744450 - -
          Affymetrix Rat Gene 1.0 ST Array 10744453 - -
          Affymetrix Rat Gene 1.0 ST Array 10744455 - -
          Affymetrix Rat Gene 1.0 ST Array 10744446 - -
          Affymetrix Rat Gene 1.0 ST Array 10744448 - -
          Affymetrix Rat Gene 1.0 ST Array 10744451 - -
          Affymetrix Rat Gene 1.0 ST Array 10744445 - -
          Affymetrix Rat Gene 1.0 ST Array 10744452 - -
          Affymetrix Rat Gene 1.1 ST Array 10744441 - -
          Rat Genome 230 2.0 Array 1376131_a_at - -
          Rat Genome 230 2.0 Array 1376131_PM_a_at - -
          Affymetrix HT Rat Focus Array 1376131_PM_a_at - -
          Affymetrix HT Rat Focus Array 1376131_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000028096.rn.1 TC1000003127.rn.1 RTA TC
          Rat Expression Set 230 1376131_PM_a_at - -
          Rat Expression Set 230 1376131_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744441 - -
          Rat Genome U34 Set rc_AI172229_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21567487 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366983 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline