Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467123_m1
          See other MKS1 GE Assays ›
          Gene Symbol:
          Mks1
          Gene Name:
          Meckel syndrome, type 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 75149782 - 75160825 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467123_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mks1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034917.2 NP_001030089.2 8 - 9 865 78
          GenBank mRNA BC099806.1 - 8 - 9 863 7938236 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001345.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mks1

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s)

          neural tube closure
          determination of left/right symmetry
          regulation of smoothened signaling pathway
          epithelial structure maintenance
          nonmotile primary cilium assembly
          cilium assembly
          embryonic digit morphogenesis
          motile cilium assembly
          embryonic skeletal system development
          branching morphogenesis of an epithelial tube
          inner ear receptor stereocilium organization
          cilium morphogenesis
          head development
          regulation of canonical Wnt signaling pathway
          common bile duct development
          regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          embryonic brain development
          regulation of Wnt signaling pathway, planar cell polarity pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P635769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129478 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P635769 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P635769 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P635769 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P635769 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81277 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10737327 - -
          Affymetrix Rat Gene 1.0 ST Array 10737329 - -
          Affymetrix Rat Gene 1.0 ST Array 10737342 - -
          Affymetrix Rat Gene 1.0 ST Array 10737330 - -
          Affymetrix Rat Gene 1.0 ST Array 10737336 - -
          Affymetrix Rat Gene 1.0 ST Array 10737340 - -
          Affymetrix Rat Gene 1.0 ST Array 10737343 - -
          Affymetrix Rat Gene 1.0 ST Array 10737338 - -
          Affymetrix Rat Gene 1.0 ST Array 10737334 - -
          Affymetrix Rat Gene 1.0 ST Array 10737331 - -
          Affymetrix Rat Gene 1.0 ST Array 10737332 - -
          Affymetrix Rat Gene 1.0 ST Array 10737339 - -
          Affymetrix Rat Gene 1.0 ST Array 10737337 - -
          Affymetrix Rat Gene 1.0 ST Array 10737335 - -
          Affymetrix Rat Gene 1.0 ST Array 10737326 - -
          Affymetrix Rat Gene 1.0 ST Array 10737341 - -
          Affymetrix Rat Gene 1.0 ST Array 10737333 - -
          Affymetrix Rat Gene 1.0 ST Array 10737328 - -
          Affymetrix Rat Gene 1.1 ST Array 10737325 - -
          Rat Genome 230 2.0 Array 1394270_PM_at - -
          Rat Genome 230 2.0 Array 1386100_PM_at - -
          Rat Genome 230 2.0 Array 1394270_at - -
          Rat Genome 230 2.0 Array 1386100_at - -
          Rat Genome 230 2.0 Array 1381355_at - -
          Rat Genome 230 2.0 Array 1381355_PM_at - -
          Affymetrix HT Rat Focus Array 1394270_PM_at - -
          Affymetrix HT Rat Focus Array 1381355_PM_at - -
          Affymetrix HT Rat Focus Array 1381355_at - -
          Affymetrix HT Rat Focus Array 1394270_at - -
          Affymetrix Rat Clariom S Assay PSR1000012393.rn.1 TC1000001345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000012392.rn.1 TC1000001345.rn.1 RTA TC
          Rat Expression Set 230 1381355_PM_at - -
          Rat Expression Set 230 1394270_at - -
          Rat Expression Set 230 1386100_at - -
          Rat Expression Set 230 1386100_PM_at - -
          Rat Expression Set 230 1394270_PM_at - -
          Rat Expression Set 230 1381355_at - -
          Affymetrix Rat Gene 1.0 ST Array 10737325 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21624372 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649920 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline