Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467133_g1
          See other DNAH2 GE Assays ›
          Gene Symbol:
          Dnah2
          Gene Name:
          dynein, axonemal, heavy chain 2
          Gene Aliases:
          DLP2, Dnahc2, axo f
          Chromosome Location:
          Chr.10: 56029022 - 56154564 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467133_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnah2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008767898.1 XP_008766120.1 37 - 38 6258 104
          XM_008767899.1 XP_008766121.1 37 - 38 6094 104
          XM_008767900.1 XP_008766122.1 31 - 32 5057 104
          GenBank mRNA AY903242.1 - 26 - 27 4157 104
          AY903243.1 - 26 - 27 4157 104
          AY903244.1 - 26 - 27 4157 104
          D26493.1 - 2 - 3 104
          U32179.1 - 1 - 2 191 104

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based movement
          metabolic process
          microtubule motor activity
          ATP binding
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P196751 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P196751 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P196751 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P196751 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P523294 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P510641 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P245583 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P412233 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15928 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P297930 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P213489 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 799 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92997 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14678 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100805 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744029 - -
          Affymetrix Rat Gene 1.0 ST Array 10744033 - -
          Affymetrix Rat Gene 1.0 ST Array 10744044 - -
          Affymetrix Rat Gene 1.0 ST Array 10744012 - -
          Affymetrix Rat Gene 1.0 ST Array 10744013 - -
          Affymetrix Rat Gene 1.0 ST Array 10743996 - -
          Affymetrix Rat Gene 1.0 ST Array 10744047 - -
          Affymetrix Rat Gene 1.0 ST Array 10743993 - -
          Affymetrix Rat Gene 1.0 ST Array 10744049 - -
          Affymetrix Rat Gene 1.0 ST Array 10744001 - -
          Affymetrix Rat Gene 1.0 ST Array 10744046 - -
          Affymetrix Rat Gene 1.0 ST Array 10744019 - -
          Affymetrix Rat Gene 1.0 ST Array 10744034 - -
          Affymetrix Rat Gene 1.0 ST Array 10744030 - -
          Affymetrix Rat Gene 1.0 ST Array 10744020 - -
          Affymetrix Rat Gene 1.0 ST Array 10744004 - -
          Affymetrix Rat Gene 1.0 ST Array 10744042 - -
          Affymetrix Rat Gene 1.0 ST Array 10744021 - -
          Affymetrix Rat Gene 1.0 ST Array 10744027 - -
          Affymetrix Rat Gene 1.0 ST Array 10744057 - -
          Affymetrix Rat Gene 1.0 ST Array 10744028 - -
          Affymetrix Rat Gene 1.0 ST Array 10744058 - -
          Affymetrix Rat Gene 1.0 ST Array 10744022 - -
          Affymetrix Rat Gene 1.0 ST Array 10744017 - -
          Affymetrix Rat Gene 1.0 ST Array 10744054 - -
          Affymetrix Rat Gene 1.0 ST Array 10744031 - -
          Affymetrix Rat Gene 1.0 ST Array 10744043 - -
          Affymetrix Rat Gene 1.0 ST Array 10744070 - -
          Affymetrix Rat Gene 1.0 ST Array 10744032 - -
          Affymetrix Rat Gene 1.0 ST Array 10744060 - -
          Affymetrix Rat Gene 1.0 ST Array 10744048 - -
          Affymetrix Rat Gene 1.0 ST Array 10744037 - -
          Affymetrix Rat Gene 1.0 ST Array 10744069 - -
          Affymetrix Rat Gene 1.0 ST Array 10744039 - -
          Affymetrix Rat Gene 1.0 ST Array 10744056 - -
          Affymetrix Rat Gene 1.0 ST Array 10744016 - -
          Affymetrix Rat Gene 1.0 ST Array 10744066 - -
          Affymetrix Rat Gene 1.0 ST Array 10744059 - -
          Affymetrix Rat Gene 1.0 ST Array 10744018 - -
          Affymetrix Rat Gene 1.0 ST Array 10744025 - -
          Affymetrix Rat Gene 1.0 ST Array 10744064 - -
          Affymetrix Rat Gene 1.0 ST Array 10744040 - -
          Affymetrix Rat Gene 1.0 ST Array 10744050 - -
          Affymetrix Rat Gene 1.0 ST Array 10744053 - -
          Affymetrix Rat Gene 1.0 ST Array 10743999 - -
          Affymetrix Rat Gene 1.0 ST Array 10744009 - -
          Affymetrix Rat Gene 1.0 ST Array 10744061 - -
          Affymetrix Rat Gene 1.0 ST Array 10744062 - -
          Affymetrix Rat Gene 1.0 ST Array 10744024 - -
          Affymetrix Rat Gene 1.0 ST Array 10743997 - -
          Affymetrix Rat Gene 1.0 ST Array 10744006 - -
          Affymetrix Rat Gene 1.0 ST Array 10744005 - -
          Affymetrix Rat Gene 1.0 ST Array 10744010 - -
          Affymetrix Rat Gene 1.0 ST Array 10744065 - -
          Affymetrix Rat Gene 1.0 ST Array 10744000 - -
          Affymetrix Rat Gene 1.0 ST Array 10744015 - -
          Affymetrix Rat Gene 1.0 ST Array 10744007 - -
          Affymetrix Rat Gene 1.0 ST Array 10743998 - -
          Affymetrix Rat Gene 1.0 ST Array 10744026 - -
          Affymetrix Rat Gene 1.0 ST Array 10744003 - -
          Affymetrix Rat Gene 1.0 ST Array 10744068 - -
          Affymetrix Rat Gene 1.0 ST Array 10743994 - -
          Affymetrix Rat Gene 1.0 ST Array 10744067 - -
          Affymetrix Rat Gene 1.0 ST Array 10744051 - -
          Affymetrix Rat Gene 1.0 ST Array 10744002 - -
          Affymetrix Rat Gene 1.0 ST Array 10744052 - -
          Affymetrix Rat Gene 1.0 ST Array 10744023 - -
          Affymetrix Rat Gene 1.0 ST Array 10744045 - -
          Affymetrix Rat Gene 1.0 ST Array 10744008 - -
          Affymetrix Rat Gene 1.0 ST Array 10743995 - -
          Affymetrix Rat Gene 1.0 ST Array 10744036 - -
          Affymetrix Rat Gene 1.0 ST Array 10744041 - -
          Affymetrix Rat Gene 1.0 ST Array 10744055 - -
          Affymetrix Rat Gene 1.0 ST Array 10744011 - -
          Affymetrix Rat Gene 1.0 ST Array 10744038 - -
          Affymetrix Rat Gene 1.0 ST Array 10744063 - -
          Affymetrix Rat Gene 1.0 ST Array 10744035 - -
          Affymetrix Rat Gene 1.0 ST Array 10744014 - -
          Affymetrix Rat Gene 1.1 ST Array 10743989 - -
          Affymetrix Rat Gene 1.0 ST Array 10743989 - -
          Rat Genome U34 Set D26493_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21294063 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline