Search
Search
| Gene Symbol: Cfap52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100968.2 | NP_001094438.2 | 8 - 9 | 1077 | 56 | |
| GenBank mRNA | BC161799.1 | - | 8 - 9 | 1077 | 7100780 | 56 |
| DQ445463.1 | - | 8 - 9 | 1089 | 56 | ||
Set Membership: |
|