Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467420_m1
          See other IFT20 GE Assays ›
          Gene Symbol:
          Ift20
          Gene Name:
          intraflagellar transport 20
          Gene Aliases:
          RGD1309400
          Chromosome Location:
          Chr.10: 65805814 - 65811353 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467420_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ift20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105815.1 NP_001099285.1 3 - 4 539 65
          XM_006246935.2 XP_006246997.1 2 - 3 353 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001170.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          smoothened signaling pathway
          visual learning
          neurogenesis
          protein localization to Golgi apparatus
          photoreceptor cell outer segment organization
          opsin transport
          intraciliary transport
          cilium assembly
          centrosome localization
          cardiac muscle cell differentiation
          regulation of canonical Wnt signaling pathway
          neural precursor cell proliferation
          protein localization to cilium
          regulation of cilium assembly
          regulation of autophagosome assembly
          opsin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004023 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004023 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004023 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1015733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10736303 - -
          Affymetrix Rat Gene 1.0 ST Array 10736301 - -
          Affymetrix Rat Gene 1.0 ST Array 10736306 - -
          Affymetrix Rat Gene 1.0 ST Array 10736305 - -
          Affymetrix Rat Gene 1.0 ST Array 10736302 - -
          Affymetrix Rat Gene 1.0 ST Array 10736304 - -
          Affymetrix Rat Gene 1.1 ST Array 10736300 - -
          Rat Genome 230 2.0 Array 1375425_PM_at - -
          Rat Genome 230 2.0 Array 1375425_at - -
          Affymetrix HT Rat Focus Array 1375425_PM_at - -
          Affymetrix HT Rat Focus Array 1375425_at - -
          Affymetrix Rat Clariom S Assay PSR1000010712.rn.1 TC1000001170.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000010713.rn.1 TC1000001170.rn.1 RTA TC
          Rat Expression Set 230 1375425_at - -
          Rat Expression Set 230 1375425_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10736300 - -
          Rat Genome U34 Set rc_AA924918_at - -
          Rat Genome U34 Set rc_AI101738_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21074714 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline