Search
Search
| Gene Symbol: Trim11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108276.1 | NP_001101746.1 | 3 - 4 | 869 | 80 | |
| XM_006246505.2 | XP_006246567.1 | 3 - 4 | 868 | 80 | ||
| GenBank mRNA | BC160894.1 | - | 3 - 4 | 819 | 7446534 | 80 |
Set Membership: |
|