Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467641_g1
          See other UQCRQ GE Assays ›
          Gene Symbol:
          Uqcrq
          Gene Name:
          ubiquinol-cytochrome c reductase, complex III subunit VII
          Gene Aliases:
          Qpc
          Chromosome Location:
          Chr.10: 38779132 - 38782487 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467641_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uqcrq
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025134.1 NP_001020305.1 1 - 2 267 65
          XM_006246316.2 XP_006246378.1 2 - 3 247 65
          GenBank mRNA AY323237.1 - 1 - 2 267 65
          FQ209456.1 - 2 - 3 188 65
          FQ209915.1 - 2 - 3 187 65
          FQ210236.1 - 2 - 3 187 65
          FQ210461.1 - 2 - 3 187 65
          FQ210609.1 - 2 - 3 187 65
          FQ210647.1 - 2 - 3 187 65
          FQ210681.1 - 2 - 3 186 65
          FQ211104.1 - 2 - 3 186 65
          FQ211208.1 - 2 - 3 187 65
          FQ211225.1 - 2 - 3 185 65
          FQ211785.1 - 2 - 3 187 65
          FQ216941.1 - 2 - 3 195 65
          FQ216945.1 - 2 - 3 186 65
          FQ217097.1 - 2 - 3 187 65
          FQ217180.1 - 2 - 3 187 65
          FQ217379.1 - 2 - 3 186 65
          FQ217445.1 - 2 - 3 196 65
          FQ217512.1 - 2 - 3 188 65
          FQ217598.1 - 2 - 3 187 65
          FQ217789.1 - 2 - 3 188 65
          FQ217857.1 - 2 - 3 189 65
          FQ217947.1 - 2 - 3 185 65
          FQ217992.1 - 2 - 3 187 65
          FQ218025.1 - 2 - 3 188 65
          FQ220995.1 - 2 - 3 187 65
          FQ222364.1 - 2 - 3 186 65
          FQ222807.1 - 2 - 3 186 65
          FQ223593.1 - 2 - 3 188 65
          FQ223655.1 - 2 - 3 186 65
          FQ223921.1 - 2 - 3 187 65
          FQ223972.1 - 2 - 3 188 65
          FQ223991.1 - 2 - 3 189 65
          FQ224065.1 - 2 - 3 188 65
          FQ224075.1 - 2 - 3 186 65
          FQ224142.1 - 2 - 3 187 65
          FQ224267.1 - 2 - 3 188 65
          FQ224342.1 - 2 - 3 196 65
          FQ224703.1 - 2 - 3 189 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Uqcrq

          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          subthalamus development
          pons development
          cerebellar Purkinje cell layer development
          hippocampus development
          thalamus development
          hypothalamus development
          pyramidal neuron development
          midbrain development
          oxidation-reduction process
          hydrogen ion transmembrane transport
          ubiquinol-cytochrome-c reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64092 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143037 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143037 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P236087 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742602 - -
          Affymetrix Rat Gene 1.0 ST Array 10742603 - -
          Affymetrix Rat Gene 1.1 ST Array 10742601 - -
          Rat Genome 230 2.0 Array 1371417_at - -
          Rat Genome 230 2.0 Array 1371417_PM_at - -
          Affymetrix HT Rat Focus Array 1371417_PM_at - -
          Affymetrix HT Rat Focus Array 1371417_at - -
          Affymetrix Rat Clariom S Assay PSR1000024441.rn.1 TC1000002784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024436.rn.1 TC1000002784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024440.rn.1 TC1000002784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024442.rn.1 TC1000002784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024438.rn.1 TC1000002784.rn.1 RTA TC
          Rat Expression Set 230 1371417_PM_at - -
          Rat Expression Set 230 1371417_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742601 - -
          Rat Genome U34 Set rc_AI233054_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22411221 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline