Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467646_m1
          See other SPDL1 GE Assays ›
          Gene Symbol:
          Spdl1
          Gene Name:
          spindle apparatus coiled-coil protein 1
          Gene Aliases:
          Ccdc99, RGD1306908
          Chromosome Location:
          Chr.10: 19628070 - 19652914 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467646_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spdl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034138.1 NP_001029310.1 11 - 12 1785 164
          XM_006246104.1 XP_006246166.1 11 - 12 1823 164
          GenBank mRNA BC105812.1 - 11 - 12 1785 7112484 164

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002532.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spdl1

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          mitotic metaphase plate congression
          protein localization to kinetochore
          cell division
          enzyme binding
          kinetochore binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697912 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86185 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062288 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062288 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062288 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P180795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36502 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75919 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742026 - -
          Affymetrix Rat Gene 1.0 ST Array 10742018 - -
          Affymetrix Rat Gene 1.0 ST Array 10742017 - -
          Affymetrix Rat Gene 1.0 ST Array 10742019 - -
          Affymetrix Rat Gene 1.0 ST Array 10742021 - -
          Affymetrix Rat Gene 1.0 ST Array 10742020 - -
          Affymetrix Rat Gene 1.0 ST Array 10742027 - -
          Affymetrix Rat Gene 1.0 ST Array 10742022 - -
          Affymetrix Rat Gene 1.0 ST Array 10742023 - -
          Affymetrix Rat Gene 1.0 ST Array 10742028 - -
          Affymetrix Rat Gene 1.0 ST Array 10742024 - -
          Affymetrix Rat Gene 1.0 ST Array 10742025 - -
          Affymetrix Rat Gene 1.1 ST Array 10742016 - -
          Rat Genome 230 2.0 Array 1382592_PM_at - -
          Rat Genome 230 2.0 Array 1382592_at - -
          Affymetrix HT Rat Focus Array 1382592_at - -
          Affymetrix HT Rat Focus Array 1382592_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000022917.rn.1 TC1000002532.rn.1 RTA TC
          Rat Expression Set 230 1382592_at - -
          Rat Expression Set 230 1382592_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742016 - -
          Rat Genome U34 Set rc_AI059054_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21127630 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline